missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-1-[2,5-dimethyl-1-(4-methylphenyl)-1H-pyrrol-3-yl]-1-ethanone, 97%, Thermo Scientific™

Product Code. 10219702
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10219702 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10219702 Supplier Thermo Scientific Maybridge Supplier No. MO07025DA
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 300557-74-8
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C15H16ClNO
MDL Number MFCD01184046
Quantity 1 g
Synonym 2-chloro-1-2,5-dimethyl-1-4-methylphenyl-1h-pyrrol-3-yl-1-ethanone, 2-chloro-1-2,5-dimethyl-1-4-methylphenyl pyrrol-3-yl ethanone, 2-chloro-1-2,5-dimethyl-1-4-methylphenyl-1h-pyrrol-3-yl ethan-1-one, 2-chloro-1-2,5-dimethyl-1-p-tolyl-1h-pyrrol-3-yl ethan-1-one, ethanone,2-chloro-1-2,5-dimethyl-1-4-methylphenyl-1h-pyrrol-3-yl, 1-2,5-dimethyl-1-4-methylphenyl pyrrol-3-yl-2-chloroethan-1-one, 2-chloro-1-2,5-dimethyl-1-p-tolyl-1h-pyrrol-3-yl ethanone, 2-chloro-1-2,5-dimethyl-1-4-methylphenyl-1h-pyrrol-3-yl ethanone
InChI Key BAEXXXPZKCRCQD-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)N2C(=CC(=C2C)C(=O)CCl)C
IUPAC Name 2-chloro-1-[2,5-dimethyl-1-(4-methylphenyl)pyrrol-3-yl]ethanone
Molecular Weight (g/mol) 261.749
PubChem CID 2794753
Formula Weight 261.75
Percent Purity 97%
Chemical Name or Material 2-Chloro-1-[2, 5-dimethyl-1-(4-methylphenyl)-1H-pyrrol-3-yl]-1-ethanone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.