missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,3-dimethylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals

Product Code. 15461036
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15461036 5 g 5g
15451036 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15461036 Supplier Thermo Scientific Alfa Aesar Supplier No. H54316.06
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 153433-26-2 | Molecular Formula: C5H10BClF4N2 | Molecular Weight: 220.40 g/mol

2-Chloro-1,3-dimethylimidazolium tetrafluoroborate is a reagent for esterification of C-terminal amino acids to Wang resin without racemization and for coupling α,α-dimethyl amino acids; and for preparation of the benzotriazol-1-yloxy derivative, a new coupling reagent. It is used as a reagent for esterification and peptide coupling of sterically hindered amino acids; preparation of efficient diazo-transfer reagents; oxidative insertion for palladium and nickel catalyst synthesis; and preparation of catalysts for use in coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-1,3-dimethylimidazolium tetrafluoroborate is a reagent for esterification of C-terminal amino acids to Wang resin without racemization and for coupling α,α-dimethyl amino acids; and for preparation of the benzotriazol-1-yloxy derivative, a new coupling reagent. It is used as a reagent for esterification and peptide coupling of sterically hindered amino acids; preparation of efficient diazo-transfer reagents; oxidative insertion for palladium and nickel catalyst synthesis; and preparation of catalysts for use in coupling reactions.

Solubility
Soluble in acetonitrile (0.1 g/ml).

Notes
Moisture and light sensitive. Incompatible with oxidizing agents and heat.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153433-26-2
Molecular Formula C5H10BClF4N2
Molecular Weight (g/mol) 220.40
MDL Number MFCD09039291
InChI Key UPLXKEIRISBKRM-UHFFFAOYSA-N
Synonym CIB
IUPAC Name 2-chloro-1,3-dimethyl-4,5-dihydro-1H-imidazol-3-ium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CN1CC[N+](C)=C1Cl

Specifications

Melting Point 175°C to 177°C
Quantity 5 g
Sensitivity Light and moisture sensitive
Solubility Information Soluble in acetonitrile (0.1g/ml).
Formula Weight 220.4
Percent Purity ≥98%
Chemical Name or Material 2-Chloro-1,3-dimethylimidazolium tetrafluoroborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.