missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,3-dinitrobenzene, 98+%

Product Code. 11321637
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11321637 5 g 5g
11311637 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11321637 Supplier Thermo Scientific Alfa Aesar Supplier No. L04593.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 606-21-3 | C6H3ClN2O4 | 202.55 g/mol

2-Chloro-1,3-dinitrobenzene can be used as intermediates of dyes and pigments. Also used as solvents in HPLC, NMR.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-1,3-dinitrobenzene can be used as intermediates of dyes and pigments. Also used as solvents in HPLC, NMR.

Solubility
Soluble in alcohol. Moderately soluble in benzene, ether. Practically insoluble in water.

Notes
Incompatible materials are oxidizing agents, Strong bases. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 606-21-3
Molecular Formula C6H3ClN2O4
Molecular Weight (g/mol) 202.55
MDL Number MFCD00014689
InChI Key BPPMIQPXQVIZNJ-UHFFFAOYSA-N
Synonym 2,6-dinitrochlorobenzene, 1-chloro-2,6-dinitrobenzene, benzene, 2-chloro-1,3-dinitro, 2-chloro-1,3-dintrobenzene, 1,3-dinitro-2-chlorobenzene, 2-chloro-1,3-dinitro-benzene, unii-lg72rnd0ab, lg72rnd0ab, 1-chloro-2,3-dinitrobenzene mixture, ambsclk-136
PubChem CID 11814
IUPAC Name 2-chloro-1,3-dinitrobenzene
SMILES [O-][N+](=O)C1=CC=CC(=C1Cl)[N+]([O-])=O

Specifications

Melting Point 83°C to 88°C
Density 1.687
Boiling Point 315°C
Quantity 5 g
UN Number UN3441
Beilstein 522187
Merck Index 14,2137
Solubility Information Soluble in alcohol. Moderately soluble in benzene,ether. Practically insoluble in water.
Formula Weight 202.55
Percent Purity ≥98%
Chemical Name or Material 2-Chloro-1,3-dinitrobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.