missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10526055
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10526055 5 g 5g
10172784 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10526055 Supplier Thermo Scientific Maybridge Supplier No. CC62901DE
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 101012-12-8
Molecular Formula C4H2ClNO2S
Molecular Weight (g/mol) 163.575
InChI Key HNJOKQPEJIWTRF-UHFFFAOYSA-N
Synonym 2-chlorothiazole-5-carboxylic acid, 2-chloro-5-thiazolecarboxylic acid, 5-thiazolecarboxylicacid, 2-chloro, 5-thiazolecarboxylic acid, 2-chloro, acmc-1brcy, 2-chlorothiazol-5-carboxylic acid, 2-chloro-5-carboxy-1,3-thiazole, 5-thiazolecarboxylicacid,2-chloro, 2-chloro-thiazole-5-carboxylic acid
PubChem CID 1481389
IUPAC Name 2-chloro-1,3-thiazole-5-carboxylic acid
SMILES C1=C(SC(=N1)Cl)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 163.58
Percent Purity 97%
Chemical Name or Material 2-Chloro-1, 3-thiazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.