missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1-methylbenzimidazole, Thermo Scientific Chemicals

Product Code. 15430319
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15430319 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15430319 Supplier Thermo Scientific Alfa Aesar Supplier No. H63559.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1849-02-1 | C8H7ClN2 | 166.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1849-02-1
Molecular Formula C8H7ClN2
Molecular Weight (g/mol) 166.61
MDL Number MFCD00769821
InChI Key UXZYKSFMGDWHGJ-UHFFFAOYSA-N
Synonym 2-chloro-1-methyl-1h-benzo d imidazole, 2-chloro-1-methyl-1h-benzimidazole, 2-chloro-1-methyl-1h-benzoimidazole, 2-chloro-1-methyl-1,3-benzodiazole, 1-methyl-1h-benzo d imidazol-2-yl chloride, 2-chloro-1-methyl-1h-1,3-benzodiazole, 1-methyl-2-chlorobenzimidazole, 2-chloro-1-methyl-benzimidazole, 1h-benzimidazole, 2-chloro-1-methyl, acmc-209em7
PubChem CID 663080
IUPAC Name 2-chloro-1-methylbenzimidazole
SMILES CN1C(Cl)=NC2=CC=CC=C12

Specifications

Color White to Yellow
Quantity 5 g
Formula Weight 166.61
Physical Form Solid
Chemical Name or Material 2-Chloro-1-methylbenzimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.