missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1-morpholinoethan-1-one, 95%, Thermo Scientific™

Product Code. 10722683
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
Product Code. Quantity unitSize
10722683 1 g 1g
10202602 10 g 10g
10326854 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 10722683

Brand: Thermo Scientific Maybridge KM10055DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1440-61-5
Molecular Formula C6H10ClNO2
Molecular Weight (g/mol) 163.601
MDL Number MFCD00721939
InChI Key YMQRPXBBBOXHNZ-UHFFFAOYSA-N
Synonym 4-2-chloroacetyl morpholine, morpholine, 4-chloroacetyl, 4-chloroacetyl morpholine, n-chloroacetyl morpholine, 2-chloro-1-morpholin-4-yl ethan-1-one, n-chloroacetylmorpholine, chloroacetic acid morpholide, 2-chloro-1-morpholinoethanone, 2-chloro-1-morpholin-4-yl-ethanone, chloroacetic acid, morpholide
PubChem CID 74040
IUPAC Name 2-chloro-1-morpholin-4-ylethanone
SMILES C1COCCN1C(=O)CCl

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 163.6
Percent Purity 95%
Chemical Name or Material 2-Chloro-1-morpholinoethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.