missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-10-[3-[[3-(dimethylamino)propyl]methylamino]propyl]phenothiazine Dihydrochloride, TRC

Product Code. 30361084
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30361084 250 mg
30359343 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30361084 Supplier Toronto Research Chemicals Supplier No. TRCC367113250MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2512203-91-5
Molecular Formula C21H28ClN3S.2ClH
Molecular Weight (g/mol) 462.91
Synonym N-[3-(2-Chloro-10H-phenothiazin-10-yl)propyl]-N,N',N'-trimethylpropane-1,3-diamine Dihydrochloride, Chlorpromazine Hydrochloride Imp. B (EP) as Dihydrochloride, 1,3-Propanediamine, N1-[3-(2-chloro-10H-phenothiazin-10-yl)propyl]-N1,N3,N3-trimethyl-, hydrochloride (1:2) (ACI)
IUPAC Name N'-[3-(2-chlorophenothiazin-10-yl)propyl]-N,N,N'-trimethylpropane-1,3-diamine;dihydrochloride
SMILES Cl.Cl.CN(C)CCCN(C)CCCN1c2ccccc2Sc3ccc(Cl)cc13

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C21H28ClN3S.2ClH/c1-23(2)12-6-13-24(3)14-7-15-25-18-8-4-5-9-20(18)26-21-11-10-17(22)16-19(21)25;;/h4-5,8-11,16H,6-7,12-15H2,1-3H3;2*1H
Formula Weight 461.1226
Chemical Name or Material N-[3-(2-Chloro-10H-phenothiazin-10-yl)propyl]-N,N',N'-trimethylpropane-1,3-diamine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.