missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-2',6'-dimethylacetanilide, 99%

Product Code. 11367805
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11367805 10 g 10g
11377805 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367805 Supplier Thermo Scientific Alfa Aesar Supplier No. L02727.09
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1131-01-7 | C10H12ClNO | 197.662 g/mol

2-Chloro-2',6'-dimethylacetanilide is used in the determination of the enantiomers of tocainide in blood plasma by gas-liquid chromatography with electron-capture detection. It is a nonsteroidal antinflammatory drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in Chloroform, Methanol. Insoluble in water.

Notes
Keep the container tightly closed in a cool, dry and well ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1131-01-7
Molecular Formula C10H12ClNO
Molecular Weight (g/mol) 197.662
MDL Number MFCD00000926
InChI Key FPQQSNUTBWFFLB-UHFFFAOYSA-N
Synonym 2-chloro-n-2,6-dimethylphenyl acetamide, 2-chloro-2',6'-dimethylacetanilide, n-chloroacetyl-2,6-dimethylaniline, 2-chloro-2',6'-acetoxylidide, chloroaceto-2,6-xylidide, acetamide, 2-chloro-n-2,6-dimethylphenyl, chloroacetamido-2,6-xylidine, 1-chloroacetylamino-2,6-dimethylbenzene, n-2,6-dimethylphenyl-2-chloroacetamide, unii-5d2b69oapx
PubChem CID 70798
IUPAC Name 2-chloro-N-(2,6-dimethylphenyl)acetamide
SMILES CC1=C(C(=CC=C1)C)NC(=O)CCl

Specifications

Melting Point 143°C to 148°C
Quantity 10 g
Beilstein 781309
Solubility Information Soluble in Chloroform,Methanol. Insoluble in water.
Formula Weight 197.67
Percent Purity 99%
Chemical Name or Material 2-Chloro-2',6'-dimethylacetanilide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.