missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-2',6'-dimethylacetanilide, TRC

Product Code. 30135343
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135343 5 g 5g
17923065 10 g 10g
17964085 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135343 Supplier Toronto Research Chemicals Supplier No. TRCC3653755G
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1131-01-7
Molecular Formula C10 H12 Cl N O
Molecular Weight (g/mol) 197.66
Synonym 2-Chloro-N-(2,6-dimethylphenyl)acetamide, Lidocaine Hydrochloride Imp. H (EP)
IUPAC Name 2-chloro-N-(2,6-dimethylphenyl)acetamide
SMILES Cc1cccc(C)c1NC(=O)CCl

Specifications

Quantity 5 g
InChI Formula InChI=1S/C10H12ClNO/c1-7-4-3-5-8(2)10(7)12-9(13)6-11/h3-5H,6H2,1-2H3,(H,12,13)
Formula Weight 197.0607
Percent Purity >95%
Chemical Name or Material 2-Chloro-N-(2,6-dimethylphenyl)acetamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.