missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-2-ethoxyacetic acid ethyl ester, 90%, remainder mainly ethyl diethoxyacetat, Thermo Scientific Chemicals

Product Code. 10476573 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10476573 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10476573 Supplier Thermo Scientific Acros Supplier No. 279870500
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34006-60-5
Molecular Formula C6H11ClO3
Molecular Weight (g/mol) 166.60
MDL Number MFCD00043953
InChI Key WDCHMODANTVRTF-UHFFFAOYNA-N
Synonym 2-chloro-2-ethoxyacetic acid ethyl ester, ethyl 2-chloro-2-ethoxy-acetate, ethyl2-chloro-2-ethoxyacetate, ethyl chloro ethoxy acetate, chloro ethoxy acetic acid ethyl ester, ethyl alpha-chloro-alpha-ethoxyacetate, ethyl alpha-chloro-alpha-ethoxy-acetate, 2-chloro-2-ethoxy-acetic acid ethyl ester, acetic acid,2-chloro-2-ethoxy-, ethyl ester
PubChem CID 2724873
IUPAC Name ethyl 2-(1-chloroethoxy)acetate
SMILES CCOC(=O)COC(C)Cl

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Brown-Orange to Colorless
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula C2H5O(Cl)CHC(O)OC2H5
Refractive Index 1.4307
Quantity 50 g
Formula Weight 166.6
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 2-Chloro-2-ethoxyacetic acid ethyl ester
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-2-ethoxyacetic acid ethyl ester, remainder mainly ethyl diethoxyacetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.