missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-2-methylpropane, 99%

Product Code. 10040610 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
2.5 L
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10040610 500 mL Glass bottle 500mL
10142470 25 mL Glass bottle 25mL
10010590 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10040610 Supplier Thermo Scientific Acros Supplier No. 109505000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

2-Chloro-2-methylpropane, 99%, C4H9Cl, CAS Number-507-20-0, 2-methyl-2-chloropropane, propane, 2-chloro-2-methyl, 2-chloroisobutane, tert-butylchloride, t-butylchloride, chlorotrimethylmethane, trimethylchloromethane, tert-butyl chloride, t-butyl chloride, 2-methyl-2-propyl chloride, 500mL | CAS: 507-20-0 | C4H9Cl | 92.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 507-20-0
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.57
MDL Number MFCD00000816
InChI Key NBRKLOOSMBRFMH-UHFFFAOYSA-N
Synonym tert-butyl chloride, propane, 2-chloro-2-methyl, tert-butylchloride, chlorotrimethylmethane, trimethylchloromethane, 2-chloroisobutane, t-butyl chloride, t-butylchloride, 2-methyl-2-chloropropane, 2-methyl-2-propyl chloride
PubChem CID 10486
IUPAC Name 2-chloro-2-methylpropane
SMILES CC(C)(C)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -25.0°C
Density 0.8500g/mL
Boiling Point 51.0°C to 52.0°C
Flash Point −27°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CCl
Refractive Index 1.3838 to 1.3858
Quantity 500 mL
Beilstein 01, 125
Fieser 02,46
Merck Index 15, 1563
Specific Gravity 0.85
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with alcohol and ether
Viscosity 0.51 mPa.s (20°C)
Formula Weight 92.57
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Chloro-2-methylpropane, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-2-methylpropane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.