missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3,4-dimethoxybenzaldehyde, 98%, ACROS Organics™

Product Code. 10009011 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
25g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10009011 25g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10009011 Supplier Thermo Scientific Acros Supplier No. 365640250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 5417-17-4
CAS Min % 97.5
CAS Max % 100.0
Melting Point 69°C to 73°C
Color Beige to Beige-Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C9H9ClO3
Linear Formula (CH3O)2C6H2(Cl)CHO
MDL Number MFCD00084942
Quantity 25 g
Synonym 2-chloroveratraldehyde, 2-chloro-3,4-dimethoxy-benzaldehyde, benzaldehyde, 2-chloro-3,4-dimethoxy, 2-chloro-3,4-dimethoxy benzaldehyde, chloroveratraldehyde, pubchem7759, acmc-1aumn, 2-chloro-3,4 dimethoxybenzaldehyde, benzaldehyde,2-chloro-3,4-dimethoxy, 2-chloro-3,4-dimethoxybenzaldehyde
InChI Key SAWHDJTZESXNMM-UHFFFAOYSA-N
SMILES COC1=C(C(=C(C=C1)C=O)Cl)OC
IUPAC Name 2-chloro-3,4-dimethoxybenzaldehyde
Molecular Weight (g/mol) 200.618
PubChem CID 79445
Formula Weight 200.62
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-3, 4-dimethoxybenzaldehyde, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.