missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3,5-dinitrobenzotrifluoride, TRC

Product Code. 30348804
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30348804 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30348804 Supplier Toronto Research Chemicals Supplier No. TRCC365680100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material 2-Chloro-3,5-dinitrobenzotrifluoride
CAS 392-95-0
Molecular Formula C7 H2 Cl F3 N2 O4
Quantity 100 g
Synonym 2-Chloro-1,5-dinitro-3-(trifluoromethyl)benzene, 2-Chloro-3,5-dinitro-α,α,α-trifluorotoluene, 1-Chloro-2,4-dinitro-6-(trifluoromethyl)benzene, 2-Chloro-α,α,α-trifluoro-3,5-dinitrotoluene
InChI Formula InChI=1S/C7H2ClF3N2O4/c8-6-4(7(9,10)11)1-3(12(14)15)2-5(6)13(16)17/h1-2H
SMILES [O-][N+](=O)c1cc(c(Cl)c(c1)C(F)(F)F)[N+](=O)[O-]
IUPAC Name 2-chloro-1,5-dinitro-3-(trifluoromethyl)benzene
Molecular Weight (g/mol) 270.55
Formula Weight 269.9655
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.