missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3,5-dinitropyridine, 98+%

Product Code. 10781165 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10781165 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10781165 Supplier Thermo Scientific Acros Supplier No. 208190050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2578-45-2 | C5H2ClN3O4 | 203.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2578-45-2
CAS Min % 98.5
CAS Max % 100.0
Melting Point 63.0°C to 66.0°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Molecular Formula C5H2ClN3O4
Linear Formula (O2H)(C5H2N)Cl
MDL Number MFCD00006233
Quantity 5 g
Beilstein 20,III,2536
Synonym pyridine, 2-chloro-3,5-dinitro, 2-chloro-3,5-dinitro-pyridine, pubchem6103, acmc-209gmh, 2-chloro3,5-dinitropyridine, 2-chloro-3,5-dinitropyridine
InChI Key QLHVJBXAQWPEDI-UHFFFAOYSA-N
SMILES C1=C(C=NC(=C1[N+](=O)[O-])Cl)[N+](=O)[O-]
IUPAC Name 2-chloro-3,5-dinitropyridine
Molecular Weight (g/mol) 203.54
PubChem CID 75738
Formula Weight 203.54
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material 2-Chloro-3,5-dinitropyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-3,5-dinitropyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.