missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-fluoropyridine, 97%, Thermo Scientific™

Product Code. 11976421 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11976421 25 g 25g
11966421 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11976421 Supplier Thermo Scientific Acros Supplier No. 442080250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 17282-04-1
Molecular Formula C5H3ClFN
Molecular Weight (g/mol) 131.54
InChI Key SVAZIMBLBHOVIR-UHFFFAOYSA-N
Synonym 3-fluoro-2-chloropyridine, 2-chloro-3-fluoro pyridine, pyridine, 2-chloro-3-fluoro, 2-chloro-3-fluoro-pyridine, pubchem2532, acmc-1cc0d, ksc174s3h, 2-chloro-3-fluoropyridine, 2-chloro-3-fluoropyridine 1g
PubChem CID 2782791
IUPAC Name 2-chloro-3-fluoropyridine
SMILES C1=CC(=C(N=C1)Cl)F

Specifications

Density 1.3230g/mL
Boiling Point 80°C (80.0 mmHg)
Flash Point 63°C
Assay Percent Range 96% min. (GC)
Quantity 25 g
Specific Gravity 1.323
Formula Weight 131.54
Percent Purity 97%
Chemical Name or Material 2-Chloro-3-fluoropyridine
compliance-icons
Product Identifier
  • 2-Chloro-3-fluoropyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.