missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-3-methyl-5-nitropyridine, 97%, Thermo Scientific™

Product Code. 10577834
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10577834 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Artikelnummer. 10577834 Leverandør Thermo Scientific Maybridge Leverandørnr. BTB09487DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Tekniske data

CAS 22280-56-4
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C6H5ClN2O2
MDL Number MFCD00173686
Quantity 1 g
Synonym 2-chloro-5-nitro-3-picoline, pyridine, 2-chloro-3-methyl-5-nitro, 2-chloro-3-methyl-5-nitro pyridine, 2-chloro-3-methyl-5-nitro-pyridine, 2-chloro-5-nitro-3-methylpyridine, pubchem10621, acmc-209fub, ksc207a2n, osioigxjuztwri-uhfffaoysa
InChI Key OSIOIGXJUZTWRI-UHFFFAOYSA-N
SMILES CC1=CC(=CN=C1Cl)[N+](=O)[O-]
IUPAC Name 2-chloro-3-methyl-5-nitropyridine
Molecular Weight (g/mol) 172.568
PubChem CID 89648
Formula Weight 172.57
Percent Purity 97%
Chemical Name or Material 2-Chloro-3-methyl-5-nitropyridine
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.