missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-3-methylquinoline, 97%, Thermo Scientific™

Product Code. 10594974
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10594974 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10594974 Supplier Thermo Scientific Maybridge Supplier No. NRB00907DA
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 57876-69-4
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H8ClN
MDL Number MFCD00100265
Quantity 1 g
Synonym 2-chloro-3-methyl-quinoline, quinoline, 2-chloro-3-methyl, zlchem 105, pubchem12431, acmc-1ayrn, methyl 2-chloroquinoline, 2-chloro-3-methylquinolin, 3-methyl-2-chloroquinoline, 2-cloro-3-methyl-quinoline, 2-chloro-3-methyl quinoline
InChI Key UMYPPRQNLXTIEQ-UHFFFAOYSA-N
SMILES CC1=CC2=CC=CC=C2N=C1Cl
IUPAC Name 2-chloro-3-methylquinoline
Molecular Weight (g/mol) 177.631
PubChem CID 339082
Formula Weight 177.63
Percent Purity 97%
Chemical Name or Material 2-chloro-3-methylquinoline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.