missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-nitro-4-picoline, 98%, Thermo Scientific™

Product Code. 10413073 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10413073 1 g Glass bottle 1g
10249863 100 mg Glass bottle 100mg
10280463 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10413073 Supplier Thermo Scientific Acros Supplier No. 379450010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 23056-39-5
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.57
MDL Number MFCD00012347
InChI Key JHARVUVBTAAPLA-UHFFFAOYSA-N
Synonym 2-chloro-3-nitro-4-picoline, pyridine, 2-chloro-4-methyl-3-nitro, 2-chloro-3-nitro-4-methylpyridine, 2-chloro-4-methyl-3-nitro-pyridine, pubchem6148, acmc-1cui4, 2-chloro-3-nitro4-picoline, ksc496c3n, 2-chlor-4-methyl-3-nitropyridin
PubChem CID 345363
IUPAC Name 2-chloro-4-methyl-3-nitropyridine
SMILES CC1=C(C(=NC=C1)Cl)[N+](=O)[O-]

Tekniske data

CAS Min % 97.5
CAS Max % 100.0
Melting Point 51°C to 53°C
Color Yellow to Beige
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 172.57
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Chloro-3-nitro-4-picoline
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • 2-Chloro-3-nitro-4-picoline
Signalord
  • Warning
Farekategori
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faresætning
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Sikkerhedssætning
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.