missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-nitropyridine, 99+%

Product Code. 10705371 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10705371 25 g Glass bottle 25g
10197563 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10705371 Supplier Thermo Scientific Acros Supplier No. 109730250
 Restricted Item
In Stock
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 5470-18-8 | C5H3ClN2O2 | 158.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 5470-18-8
Molecular Formula C5H3ClN2O2
Molecular Weight (g/mol) 158.54
MDL Number MFCD00006232
InChI Key UUOLETYDNTVQDY-UHFFFAOYSA-N
Synonym pyridine, 2-chloro-3-nitro, 3-nitro-2-chloropyridine, 2-chloro-3-nitro-pyridine, 2-chloro-3-nitro pyridine, 2-chlor-3-nitropyridin, zlchem 303, pubchem1229, 2-chloro-nitropyridine, 2-chloro-3nitropyridine, 2-chloro3-nitropyridine
PubChem CID 79613
IUPAC Name 2-chloro-3-nitropyridine
SMILES C1=CC(=C(N=C1)Cl)[N+](=O)[O-]

Specifica

CAS Min % 99.0
CAS Max % 100.0
Melting Point 100.0°C to 103.0°C
Color Yellow
Flash Point 185°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Linear Formula Cl(C5H3N)NO2
Refractive Index 1.4540 to 1.4560
Quantity 25 g
Beilstein 20, I, 82
Solubility Information Solubility in water: insoluble
Formula Weight 158.54
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 2-chloro-3-nitropyridine
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • 2-Chloro-3-nitropyridine
Avvertenza
  • Warning
Categoria di pericolo
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Consigli di prudenza
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.