missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-quinolinecarbonyl chloride, Tech., Thermo Scientific™

Product Code. 10575483
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10575483 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10575483 Supplier Thermo Scientific Maybridge Supplier No. MO01102DA
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 136812-19-6
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C10H5Cl2NO
MDL Number MFCD06200869
Quantity 1 g
Synonym 2-chloro-3-quinolinecarbonyl chloride, 2-chloro-3-quinolinecarbonylchloride
InChI Key QZUMFCDPDBLWBV-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C=C(C(=N2)Cl)C(=O)Cl
IUPAC Name 2-chloroquinoline-3-carbonyl chloride
Molecular Weight (g/mol) 226.056
PubChem CID 2794665
Formula Weight 226.06
Percent Purity 95%
Grade Technical
Chemical Name or Material 2-Chloro-3-quinolinecarbonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.