missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4-fluoro-5-nitrobenzoic Acid, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | 2-Chloro-4-fluoro-5-nitrobenzoic Acid |
| CAS | 114776-15-7 |
| Molecular Formula | C7H3ClFNO4 |
| Quantity | 250 mg |
| Synonym | 2-Chloro-4-fluoro-5-nitro-benzoic Acid |
| InChI Formula | InChI=1S/C7H3ClFNO4/c8-4-2-5(9)6(10(13)14)1-3(4)7(11)12/h1-2H,(H,11,12) |
| SMILES | OC(=O)c1cc(c(F)cc1Cl)[N+](=O)[O-] |
| IUPAC Name | 2-chloro-4-fluoro-5-nitrobenzoic acid |
| Molecular Weight (g/mol) | 219.55 |
| Formula Weight | 218.9735 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction