missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-4-fluorobenzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10317214
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10317214 10 g 10g
10184194 1 g 1g
10002034 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10317214 Supplier Thermo Scientific Maybridge Supplier No. TL00318EA
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 85958-57-2
Molecular Formula C6H3Cl2FO2S
Molecular Weight (g/mol) 229.046
MDL Number MFCD00052957
InChI Key FCFPJKHVCHKCMP-UHFFFAOYSA-N
Synonym 2-chloro-4-fluorobenzene-1-sulfonyl chloride, 2-chloro-4-fluorobenzenesulphonyl chloride, benzenesulfonyl chloride, 2-chloro-4-fluoro, benzenesulfonyl chloride, 2-chloro-4-fluoro-9ci, pubchem5092, acmc-209wex, buttpark 9457-18, 2-chloro-4-fluorobenzene-1-sulfonylchloride, ksc447s0p, chloro 2-chloro-4-fluorophenyl sulfone
PubChem CID 522846
IUPAC Name 2-chloro-4-fluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1F)Cl)S(=O)(=O)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula Cl(F)C6H3(SO2Cl)
Quantity 10 g
Formula Weight 229.06
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-fluorobenzene-1-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.