missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorobenzenesulfonyl chloride, 98%, Thermo Scientific™

Product Code. 11438813
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11438813

Brand: Thermo Scientific Alfa Aesar A10536.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85958-57-2
Molecular Formula C6H3Cl2FO2S
Molecular Weight (g/mol) 229.046
MDL Number MFCD00052957
InChI Key FCFPJKHVCHKCMP-UHFFFAOYSA-N
Synonym 2-chloro-4-fluorobenzene-1-sulfonyl chloride, 2-chloro-4-fluorobenzenesulphonyl chloride, benzenesulfonyl chloride, 2-chloro-4-fluoro, benzenesulfonyl chloride, 2-chloro-4-fluoro-9ci, pubchem5092, acmc-209wex, buttpark 9457-18, 2-chloro-4-fluorobenzene-1-sulfonylchloride, ksc447s0p, chloro 2-chloro-4-fluorophenyl sulfone
PubChem CID 522846
IUPAC Name 2-chloro-4-fluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1F)Cl)S(=O)(=O)Cl

Specifications

Melting Point 28°C to 31°C
Density 1.57
Boiling Point 243°C to 244°C
Flash Point >110°C (230°F)
Refractive Index 1.555
Quantity 1 g
UN Number UN3265
Beilstein 5520034
Sensitivity Moisture sensitive
Formula Weight 229.06
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-fluorobenzenesulfonyl chloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.