missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methyl-3-nitropyridine, TRC

Product Code. 30359529
Change view
Click to view available options
Quantity:
500 mg
1 g
2 g
Unit Size:
1g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359529 2 g 2g
30363331 500 mg -
30362203 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30359529 Supplier Toronto Research Chemicals Supplier No. TRCC3691452G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23056-39-5
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.57
Synonym 2-Chloro-3-nitro-4-picoline, 2-Chloro-3-nitro-4-methylpyridine, NSC 402977
IUPAC Name 2-chloro-4-methyl-3-nitropyridine
SMILES Cc1ccnc(Cl)c1[N+](=O)[O-]

Specifications

Quantity 2 g
InChI Formula InChI=1S/C6H5ClN2O2/c1-4-2-3-8-6(7)5(4)9(10)11/h2-3H,1H3
Formula Weight 172.004
Chemical Name or Material 2-Chloro-4-methyl-3-nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.