missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methyl-3-nitropyridine, TRC

Product Code. 30363331
Change view
Click to view available options
Quantity:
500 mg
1 g
2 g
Unit Size:
1g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30363331 500 mg -
30359529 2 g 2g
30362203 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30363331 Supplier Toronto Research Chemicals Supplier No. TRCC369145500MG
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23056-39-5
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.57
Synonym 2-Chloro-3-nitro-4-picoline, 2-Chloro-3-nitro-4-methylpyridine, NSC 402977
IUPAC Name 2-chloro-4-methyl-3-nitropyridine
SMILES Cc1ccnc(Cl)c1[N+](=O)[O-]

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C6H5ClN2O2/c1-4-2-3-8-6(7)5(4)9(10)11/h2-3H,1H3
Formula Weight 172.004
Chemical Name or Material 2-Chloro-4-methyl-3-nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.