missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methylnicotinamide, TRC

Product Code. 30364723
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30364723 50 mg 50mg
30363782 10 mg 10mg
30356404 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30364723 Supplier Toronto Research Chemicals Supplier No. TRCC37424550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152362-01-1
Molecular Formula C7H7N2OCl
Molecular Weight (g/mol) 170.6
Synonym 2-Chloro-4-methyl-3-pyridinecarboxamide, 2-Chloro-4-methyl-nicotinamide
IUPAC Name 2-chloro-4-methylpyridine-3-carboxamide
SMILES Cc1ccnc(Cl)c1C(=O)N

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C7H7ClN2O/c1-4-2-3-10-6(8)5(4)7(9)11/h2-3H,1H3,(H2,9,11)
Formula Weight 170.0247
Chemical Name or Material 2-Chloro-4-methylnicotinamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.