missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-nitroaniline, 98+%

Product Code. 11457917
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11457917 5000 g 5000g
11447917 250 g 250g
11437917 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11457917 Supplier Thermo Scientific Alfa Aesar Supplier No. A16586.0I
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 121-87-9 | C6H5ClN2O2 | 172.568 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-87-9
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.568
MDL Number MFCD00007665
InChI Key LOCWBQIWHWIRGN-UHFFFAOYSA-N
Synonym ocpna, 2-chloro-4-nitrobenzenamine, o-chloro-p-nitroaniline, 4-nitro-2-chloroaniline, benzenamine, 2-chloro-4-nitro, 1-amino-2-chloro-4-nitrobenzene, aniline, 2-chloro-4-nitro, unii-xsa3zx337b, 2-chloro-4-nitro-phenylamine, ortho-chloro-para-nitroaniline
PubChem CID 8492
IUPAC Name 2-chloro-4-nitroaniline
SMILES C1=CC(=C(C=C1[N+](=O)[O-])Cl)N

Specifications

Melting Point 106°C to 109°C
Boiling Point 320°C (decomposition)
Flash Point 193°C (379°F)
Odor Weak, Characteristic
Quantity 5000 g
UN Number UN2237
Beilstein 638657
Formula Weight 172.57
Percent Purity ≥98%
Chemical Name or Material 2-Chloro-4-nitroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.