missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-cyanopyridine, 97%

Product Code. 11440863
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440863 1 g 1g
11450863 5 g 5g
11460863 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440863 Supplier Thermo Scientific Alfa Aesar Supplier No. A13291.03
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 33252-28-7 | C6H3ClN2 | 138.554 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33252-28-7
Molecular Formula C6H3ClN2
Molecular Weight (g/mol) 138.554
MDL Number MFCD00084941
InChI Key ORIQLMBUPMABDV-UHFFFAOYSA-N
Synonym 2-chloro-5-cyanopyridine, 6-chloronicotinonitrile, 2-chloropyridine-5-carbonitrile, 6-chloro-3-pyridinecarbonitrile, 6-chloronicotinitrile, 6-chloronicotinontrile, 6-chloro-nicotinonitrile, 2-chloro-5-cyano-pyridine, 3-pyridinecarbonitrile, 6-chloro, 6-chloro nicotinonitrile
PubChem CID 5152094
IUPAC Name 6-chloropyridine-3-carbonitrile
SMILES C1=CC(=NC=C1C#N)Cl

Specifications

Melting Point 116°C to 119°C
Boiling Point 105°C to 107°C (1 mmHg)
Quantity 1 g
UN Number UN3439
Beilstein 113867
Formula Weight 138.56
Percent Purity 98%
Chemical Name or Material 2-Chloro-5-cyanopyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.