missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-5-ethylpyrimidine, 97%, Thermo Scientific™

Product Code. 11916551 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11916551 5 g Glass bottle 5g
11906551 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11916551 Supplier Thermo Scientific Acros Supplier No. 441730050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 111196-81-7
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.59
MDL Number MFCD00799503
InChI Key BGLLZQRUXJGTAD-UHFFFAOYSA-N
Synonym 5-ethyl-2-chloropyrimidine, 2-chloro-5-ethyl-pyrimidine, pyrimidine, 2-chloro-5-ethyl, 2-chloro-5-ethyl pyrimidine, pubchem7032, acmc-1brw8, 2-chloro-5-ethylpyrimdine, 2-chloro-5-ethylpyrimidin, 2-chloro-5-ethylpyrimidine, ksc493s1b
PubChem CID 3572763
IUPAC Name 2-chloro-5-ethylpyrimidine
SMILES CCC1=CN=C(Cl)N=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1740g/mL
Boiling Point 223°C
Flash Point >109°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 1.174
Formula Weight 142.59
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 2-chloro-5-ethylpyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-5-ethylpyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.