missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-ethylpyrimidine, 98%

Product Code. 11327029
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327029 5 g 5g
11317029 1 g 1g
11337029 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11327029 Supplier Thermo Scientific Alfa Aesar Supplier No. H26856.06
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 111196-81-7 | C6H7ClN2 | 142.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111196-81-7
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.59
MDL Number MFCD00799503
InChI Key BGLLZQRUXJGTAD-UHFFFAOYSA-N
Synonym 5-ethyl-2-chloropyrimidine, 2-chloro-5-ethyl-pyrimidine, pyrimidine, 2-chloro-5-ethyl, 2-chloro-5-ethyl pyrimidine, pubchem7032, acmc-1brw8, 2-chloro-5-ethylpyrimdine, 2-chloro-5-ethylpyrimidin, 2-chloro-5-ethylpyrimidine, ksc493s1b
PubChem CID 3572763
IUPAC Name 2-chloro-5-ethylpyrimidine
SMILES CCC1=CN=C(Cl)N=C1

Specifications

Density 1.174
Boiling Point 223°C
Flash Point >110°C (230°F)
Refractive Index 1.521
Quantity 5 g
Sensitivity Moisture sensitive; May form a precipitate
Formula Weight 142.59
Percent Purity 98%
Chemical Name or Material 2-Chloro-5-ethylpyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.