missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-5-methylphenol, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 2-Chloro-5-methylphenol |
| CAS | 615-74-7 |
| Molecular Formula | C7 H7 Cl O |
| Quantity | 5 g |
| Synonym | m-Cresol, 6-chloro- (7CI,8CI), 2-Chloro-5-methylphenol, 1-Chloro-2-hydroxy-4-methylbenzene, 3-Methyl-6-chlorophenol, 4-Chloro-3-hydroxytoluene, 5-Methyl-2-chlorophenol, 6-Chloro-3-methylphenol, 6-Chloro-m-cresol |
| InChI Formula | InChI=1S/C7H7ClO/c1-5-2-3-6(8)7(9)4-5/h2-4,9H,1H3 |
| SMILES | Cc1ccc(Cl)c(O)c1 |
| IUPAC Name | 2-chloro-5-methylphenol |
| Molecular Weight (g/mol) | 142.58 |
| Formula Weight | 142.0185 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction