missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-methylpyridine, 97%

Product Code. 15435368
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15435368 5 mL 5mL
15445368 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15435368 Supplier Thermo Scientific Alfa Aesar Supplier No. H59145.AA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 18368-64-4 | C6H6ClN | 127.57 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18368-64-4
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.57
MDL Number MFCD00792460
InChI Key VXLYOURCUVQYLN-UHFFFAOYSA-N
Synonym 2-chloro-5-picoline, 6-chloro-3-picoline, pyridine, 2-chloro-5-methyl, 2-chloro-5-methyl-pyridine, 2-chloro-5-methyl pyridine, 3-picoline, 6-chloro, pubchem1193, acmc-1bvpj, 5-methyl-2-chloropyridine
PubChem CID 581393
IUPAC Name 2-chloro-5-methylpyridine
SMILES CC1=CC=C(Cl)N=C1

Specifications

Density 1.169
Boiling Point 97°C (30 mmHg)
Refractive Index 1.535
Quantity 5 mL
Solubility Information Slightly soluble in water.
Formula Weight 127.57
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-methylpyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.