missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-methylpyrimidine, 97%

Product Code. 15452119
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15452119 5 g 5g
15442119 1 g 1g
15462119 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15452119 Supplier Thermo Scientific Alfa Aesar Supplier No. H64773.06
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 22536-61-4 | C5H5ClN2 | 128.559 g/mol

Used in organic synthesis, catalytic agent, petrochemical additive and synthetic chemistry. Also used as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in organic synthesis, catalytic agent, petrochemical additive and synthetic chemistry. Also used as intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22536-61-4
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.559
MDL Number MFCD09260903
InChI Key APRMCBSTMFKLEI-UHFFFAOYSA-N
Synonym pyrimidine, 2-chloro-5-methyl, 2-chloro-5-methyl-pyrimidine, pubchem11095, 2-chloro-5-methyl pyrimidine, ksc497i7j
PubChem CID 581719
IUPAC Name 2-chloro-5-methylpyrimidine
SMILES CC1=CN=C(N=C1)Cl

Specifications

Melting Point 89°C to 92°C
Quantity 5 g
Beilstein 108732
Solubility Information Slightly soluble in water.
Formula Weight 128.56
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Chloro-5-methylpyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.