missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-methylthiophene, 97%

Product Code. 11430227
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430227 25 g 25g
11420227 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430227 Supplier Thermo Scientific Alfa Aesar Supplier No. B21657.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17249-82-0 | C5H5ClS | 132.605 g/mol

It is used in the synthesis of dithienylcyclopentene optical molecular switches. It is also used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of dithienylcyclopentene optical molecular switches. It is also used as an active pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix with water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 17249-82-0
Molecular Formula C5H5ClS
Molecular Weight (g/mol) 132.605
MDL Number MFCD01632145
InChI Key JSMMZMYGEVUURX-UHFFFAOYSA-N
Synonym thiophene, 2-chloro-5-methyl, pubchem5197, acmc-209e4p, 2-chloro-5-methyl-thiophene, ksc497i0h, 2-chloranyl-5-methyl-thiophene
PubChem CID 140208
IUPAC Name 2-chloro-5-methylthiophene
SMILES CC1=CC=C(S1)Cl

Tekniske data

Density 1.21
Boiling Point 155°C to 156°C
Flash Point 47°C (117°F)
Odor Characteristic
Refractive Index 1.5356
Quantity 25 g
UN Number UN1992
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 132.61
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-methylthiophene
Vis mere Vis mindre

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.