missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-n-pentylpyrimidine, 98%

Product Code. 11343019
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343019 1 g 1g
11353019 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11343019 Supplier Thermo Scientific Alfa Aesar Supplier No. H27808.03
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 154466-62-3 | C9H13ClN2 | 184.667 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154466-62-3
Molecular Formula C9H13ClN2
Molecular Weight (g/mol) 184.667
MDL Number MFCD04038093
InChI Key WMARUIMXVALOMF-UHFFFAOYSA-N
Synonym 2-chloro-5-n-pentylpyrimidine, pubchem9038, 5-pentyl-2-chloropyrimidine, 2-chloro-5-pentyl pyrimidine, 2-chloranyl-5-pentyl-pyrimidine
PubChem CID 16756526
IUPAC Name 2-chloro-5-pentylpyrimidine
SMILES CCCCCC1=CN=C(N=C1)Cl

Specifications

Density 1.085
Boiling Point 124°C (3 mmHg)
Flash Point 97°C (207°F)
Refractive Index 1.504
Quantity 1 g
Formula Weight 184.67
Percent Purity 98%
Chemical Name or Material 2-Chloro-5-n-pentylpyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.