missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-n-propylpyrimidine, 98%, Thermo Scientific™

Product Code. 11387219
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11387219 10 g 10g
11377219 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387219 Supplier Thermo Scientific Alfa Aesar Supplier No. H27409.09
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 219555-98-3
Molecular Formula C7H9ClN2
Molecular Weight (g/mol) 156.61
MDL Number MFCD00799403
InChI Key WVPHBBSAPVBUGZ-UHFFFAOYSA-N
Synonym 2-chloro-5-n-propylpyrimidine, 2-chloro-5-propyl-pyrimidine, 2-chloro-5-n-propyl-pyrimidine, pubchem7036, acmc-1ccry, 5-propyl-2-chloropyrimidine, 2-chloro5-n-propylpyrimidine, 2-chloranyl-5-propyl-pyrimidine
PubChem CID 2735784
IUPAC Name 2-chloro-5-propylpyrimidine
SMILES CCCC1=CN=C(Cl)N=C1

Specifications

Density 1.142
Boiling Point 100°C (3 mmHg)
Refractive Index 1.514
Quantity 10 g
Formula Weight 156.62
Percent Purity 98%
Chemical Name or Material 2-Chloro-5-n-propylpyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.