missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitroaniline, 98%

Product Code. 11417417
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11417417 500 g 500g
11407417 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11417417 Supplier Thermo Scientific Alfa Aesar Supplier No. A15102.36
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6283-25-6 | C6H5ClN2O2 | 172.568 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6283-25-6
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.568
MDL Number MFCD00007668
InChI Key KWIXNFOTNVKIGM-UHFFFAOYSA-N
Synonym benzenamine, 2-chloro-5-nitro, 6-chloro-3-nitroaniline, 2-chloro-5-nitro aniline, aniline, 2-chloro-5-nitro, 2-chloro-5-nitrobenzenamine, aniline, 6-chloro-3-nitro, 2-chloro-5-nitrophenylamine, 2-chloro-5-nitro-benzamine, pubchem21904, 3-nitro-6-chloroaniline
PubChem CID 22691
IUPAC Name 2-chloro-5-nitroaniline
SMILES C1=CC(=C(C=C1[N+](=O)[O-])N)Cl

Specifications

Melting Point 117°C to 121°C
Flash Point 191°C (375°F)
Odor Weak, Characteristic
Quantity 500 g
UN Number UN2237
Beilstein 2208878
Formula Weight 172.57
Percent Purity 98%
Chemical Name or Material 2-Chloro-5-nitroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.