missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-picoline, 94%, Thermo Scientific™

Product Code. 10534921 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10534921 25 g Glass bottle 25g
10524921 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10534921 Supplier Thermo Scientific Acros Supplier No. 344030250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 18368-64-4
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.57
MDL Number MFCD00792460
InChI Key VXLYOURCUVQYLN-UHFFFAOYSA-N
Synonym 2-chloro-5-picoline, 6-chloro-3-picoline, pyridine, 2-chloro-5-methyl, 2-chloro-5-methyl-pyridine, 2-chloro-5-methyl pyridine, 3-picoline, 6-chloro, pubchem1193, acmc-1bvpj, 5-methyl-2-chloropyridine
PubChem CID 581393
IUPAC Name 2-chloro-5-methylpyridine
SMILES CC1=CC=C(Cl)N=C1

Specifications

CAS Min % 92.0
CAS Max % 100.0
Color Orange-Pink or Straw-Yellow to Yellow
Density 1.1690g/mL
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (GC)
Packaging Glass bottle
Refractive Index 1.5260 to 1.53
Quantity 25 g
Specific Gravity 1.169
Formula Weight 127.57
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material 2-Chloro-5-picoline
Show More Show Less
compliance-icons
Product Identifier
  • 2-chloro-5-methylpyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.