missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(trifluoromethyl)pyrazine, 97%

Product Code. 11916771 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11916771 5 g 5g
11906771 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11916771 Supplier Thermo Scientific Acros Supplier No. 446480050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

CAS: 799557-87-2 | C5H2ClF3N2 | 182.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 799557-87-2
Molecular Formula C5H2ClF3N2
Molecular Weight (g/mol) 182.53
InChI Key AIEGIFIEQXZBCP-UHFFFAOYSA-N
Synonym 2-chloro-5-trifluoromethyl pyrazine, 5-chloro-2-trifluoromethylpyrazine, pyrazine, 2-chloro-5-trifluoromethyl, 2-chloro-5-trifluoromethyl-pyrazine
PubChem CID 18987645
IUPAC Name 2-chloro-5-(trifluoromethyl)pyrazine
SMILES C1=C(N=CC(=N1)Cl)C(F)(F)F

Specifications

Boiling Point 88°C (118.0 mmHg)
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 182.53
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)pyrazine
compliance-icons compliance-icons
Product Identifier
  • 2-Chloro-5-(trifluoromethyl)pyrazine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.