missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-6-methoxy-3-nitropyridine, 97%, Thermo Scientific™

Product Code. 10782313
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10782313 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10782313 Supplier Thermo Scientific Maybridge Supplier No. RJC01057EA
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 38533-61-8
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C6H5ClN2O3
MDL Number MFCD00130268
Quantity 10 g
Synonym 2-chloro-3-nitro-6-methoxypyridine, pyridine, 2-chloro-6-methoxy-3-nitro, 2-chloro-6-methoxy-3-nitro-pyridine, 2-chloro-6-methoxy-3 or 5-nitropyridine, pubchem4044, acmc-1agmz, ksc226c7t, 2-chloro-3nitro-6-methoxy-pyridine, 6-methoxy-2-chloro-3-nitropyridine, 2-chloro-6-methoxy-3-nitropyridine
InChI Key DVRGUTNVDGIKTP-UHFFFAOYSA-N
SMILES COC1=CC=C(C(Cl)=N1)[N+]([O-])=O
IUPAC Name 2-chloro-6-methoxy-3-nitropyridine
Molecular Weight (g/mol) 188.57
PubChem CID 2795029
Formula Weight 188.57
Percent Purity 97%
Chemical Name or Material 2-chloro-6-methoxy-3-nitropyridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.