missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methoxy-7-deazapurine, TRC

Product Code. 30353930
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30353930 100 mg 100mg
30367170 500 mg 500mg
30362022 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30353930 Supplier Toronto Research Chemicals Supplier No. TRCC367950100MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96022-77-4
Molecular Formula C7 H6 Cl N3 O
Molecular Weight (g/mol) 183.6
IUPAC Name 2-chloro-4-methoxy-3H-pyrrolo[2,3-d]pyrimidine
SMILES COc1[nH]c(Cl)nc2nccc12

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C7H6ClN3O/c1-12-6-4-2-3-9-5(4)10-7(8)11-6/h2-3H,1H3,(H,9,10,11)
Formula Weight 183.0199
Chemical Name or Material 2-Chloro-6-methoxy-7-deazapurine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.