missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methoxy-7-deazapurine, TRC

Product Code. 30367170
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367170 500 mg 500mg
30353930 100 mg 100mg
30362022 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30367170 Supplier Toronto Research Chemicals Supplier No. TRCC367950500MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96022-77-4
Molecular Formula C7 H6 Cl N3 O
Molecular Weight (g/mol) 183.6
IUPAC Name 2-chloro-4-methoxy-3H-pyrrolo[2,3-d]pyrimidine
SMILES COc1[nH]c(Cl)nc2nccc12

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C7H6ClN3O/c1-12-6-4-2-3-9-5(4)10-7(8)11-6/h2-3H,1H3,(H,9,10,11)
Formula Weight 183.0199
Chemical Name or Material 2-Chloro-6-methoxy-7-deazapurine
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.