missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-6-methyl-4-(trifluoromethyl)pyridine, 97%, Thermo Scientific™

Product Code. 10732863
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10732863 10 g 10g
10574695 5 g 5g
10634965 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10732863 Supplier Thermo Scientific Maybridge Supplier No. CD10452EA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 22123-14-4
Molecular Formula C7H5ClF3N
Molecular Weight (g/mol) 195.57
MDL Number MFCD00052900
InChI Key SXLBWNSGCIEART-UHFFFAOYSA-N
Synonym 2-chloro-6-methyl-4-trifluoromethyl pyridine, 2-chloro-6-methyl-4-trifluoromethyl-pyridine, 2-chlor-6-methyl-4-trifluormethyl pyridine, 2-chloro-4-trifluoromethyl-6-methylpyridine, pyridine, 2-chloro-6-methyl-4-trifluoromethyl, pubchem9219, 6-chloro-4-trifluoromethyl-2-picoline, 2-chloranyl-6-methyl-4-trifluoromethyl pyridine
PubChem CID 2736626
IUPAC Name 2-chloro-6-methyl-4-(trifluoromethyl)pyridine
SMILES CC1=NC(Cl)=CC(=C1)C(F)(F)F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 195.57
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-methyl-4-(trifluoromethyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.