missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylphenyl isocyanate, 97%

Product Code. 11342547
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11342547 5 g 5g
11332547 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342547 Supplier Thermo Scientific Alfa Aesar Supplier No. L11645.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 40398-01-4 | C8H6ClNO | 167.592 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40398-01-4
Molecular Formula C8H6ClNO
Molecular Weight (g/mol) 167.592
MDL Number MFCD00037030
InChI Key FBTQQNYGMICJQZ-UHFFFAOYSA-N
Synonym 2-chloro-6-methylphenyl isocyanate, 2-chloro-6-methylphenylisocyanate, benzene, 1-chloro-2-isocyanato-3-methyl, acmc-1ake5, 2-chloro-6-methylbenzenisocyanate, 2-chloro 6-methyl phenylisocyanate, 2-chloro 6-methyl phenyl isocyanate, 2-chloro-1-isocyanato-6-methyl benzene, 1-chloro-2-isocyanato-3-methyl-benzene, 6-chloro-2-methylphenylisocyanate
PubChem CID 268123
IUPAC Name 1-chloro-2-isocyanato-3-methylbenzene
SMILES CC1=C(C(=CC=C1)Cl)N=C=O

Specifications

Density 1.231
Quantity 5 g
Refractive Index 1.557
UN Number UN2206
Beilstein 2690768
Boiling Point 79°C to 80°C (2 mmHg)
Formula Weight 167.6
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-methylphenyl isocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.