missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-6-methylpyrazine, 97%

Product Code. 15470449
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15470449 5 g 5g
15460449 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15470449 Supplier Alfa Aesar™ Supplier No. H63650.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 38557-71-0
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.56
MDL Number MFCD00055032
InChI Key CKUVSPQGYLELRG-UHFFFAOYSA-N
Synonym 2-chloro-6-methyl-pyrazine, pyrazine, 2-chloro-6-methyl, 6-chloro-2-methylpyrazine, 2-chloro-6-methyl-1,4-diazine, 2-methyl-6-chloropyrazine, pubchem18034, acmc-1afqe, 2-chloro-6 methylpyrazine, ksc497m1j
PubChem CID 7170095
IUPAC Name 2-chloro-6-methylpyrazine
SMILES CC1=CN=CC(Cl)=N1

Specifications

Quantity 5 g
Formula Weight 128.56
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Chloro-6-methylpyrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.