missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylpyrazine, 97%, Thermo Scientific™

Product Code. 10505303
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10505303 1 g Amber Glass Bottle 1g
10482983 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10505303 Supplier Thermo Scientific Maybridge Supplier No. MO07699DA
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 38557-71-0
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.56
MDL Number MFCD00055032
InChI Key CKUVSPQGYLELRG-UHFFFAOYSA-N
Synonym 2-chloro-6-methyl-pyrazine, pyrazine, 2-chloro-6-methyl, 6-chloro-2-methylpyrazine, 2-chloro-6-methyl-1,4-diazine, 2-methyl-6-chloropyrazine, pubchem18034, acmc-1afqe, 2-chloro-6 methylpyrazine, ksc497m1j
PubChem CID 7170095
IUPAC Name 2-chloro-6-methylpyrazine
SMILES CC1=CN=CC(Cl)=N1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 128.56
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-methylpyrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.