missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-8-cyclopentyl-5-methyl-8H-pyrido[2,3-d]pyrimidin-7-one, 97%

Product Code. 15461096
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15461096 250 mg 250mg
15441096 1 g 1g
15451096 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15461096 Supplier Thermo Scientific Alfa Aesar Supplier No. H54363.MD
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1013916-37-4 | C13H14ClN3O | 263.73 g/mol

2-Chloro-8-cyclopentyl-5-methyl-8H-pyrido[2,3-d]pyrimidin-7-one is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-8-cyclopentyl-5-methyl-8H-pyrido[2,3-d]pyrimidin-7-one is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1013916-37-4
Molecular Formula C13H14ClN3O
Molecular Weight (g/mol) 263.73
MDL Number MFCD13181207
InChI Key BSKNQSYIDZUXQT-UHFFFAOYSA-N
PubChem CID 44248248
IUPAC Name 2-chloro-8-cyclopentyl-5-methylpyrido[2,3-d]pyrimidin-7-one
SMILES CC1=CC(=O)N(C2CCCC2)C2=NC(Cl)=NC=C12

Specifications

Quantity 250 mg
Beilstein 15642863
Synonym 2-chloro-8-cyclopentyl-5-methylpyrido 2,3-d pyrimidin-7 8h-one, 2-chloro-8-cyclopentyl-5-methyl-8h-pyrido 2,3-d pyrimidin-7-one, 2-chloro-8-cyclopentyl-5-methyl-7h,8h-pyrido 2,3-d pyrimidin-7-one, 2-chloro-8-cyclopentyl-5-methylpyrido 2,3-d pyrimidin-7-one, 2-chloro-8-cyclopentyl-5-methyl-8h-pyrido 2,3-d py, pyrido 2,3-d pyrimidin-7 8h-one,2-chloro-8-cyclopentyl-5-methyl, pyrido 2,3-d pyrimidin-7 8h-one, 2-chloro-8-cyclopentyl-5-methyl
Solubility Information Slightly soluble in water.
Formula Weight 263.72
Percent Purity 97%
Chemical Name or Material 2-Chloro-8-cyclopentyl-5-methyl-8H-pyrido[2,3-d]pyrimidin-7-one

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.