missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro Adenosine, TRC

Product Code. 30132894
Change view
Click to view available options
Quantity:
10 mg
50 mg
25 g
Unit Size:
10mg
25g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132894 50 mg 50mg
17947634 25 g 25g
17909924 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132894 Supplier Toronto Research Chemicals Supplier No. TRCC36400050MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146-77-0
Molecular Formula C10 H12 Cl N5 O4
Molecular Weight (g/mol) 301.69
Synonym Adenosine, 2-chloro-, 2-Chloroadenosine, 2-Chloro-D-adenosine, Antibiotic AT 265B, CADO, NSC 36896
IUPAC Name (2R,3R,4S,5R)-2-(6-amino-2-chloropurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES Nc1nc(Cl)nc2c1ncn2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H12ClN5O4/c11-10-14-7(12)4-8(15-10)16(2-13-4)9-6(19)5(18)3(1-17)20-9/h2-3,5-6,9,17-19H,1H2,(H2,12,14,15)/t3-,5-,6-,9-/m1/s1
Formula Weight 301.0578
Chemical Name or Material 2-Chloro Adenosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.