missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-L-phenylalanine, 98+%

Product Code. 15463617
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Tamaño de la unidad
15463617 250 mg 250mg
11486240 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15463617 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor H27135.MD
 Restricted Item
In Stock
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 103616-89-3 | C9H10ClNO2 | 199.63 g/mol

The largest gain in affinity of all tested unnatural amino acids was observed for 2-chloro-L-phenylalanine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The largest gain in affinity of all tested unnatural amino acids was observed for 2-chloro-L-phenylalanine.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 103616-89-3
Molecular Formula C9H10ClNO2
Molecular Weight (g/mol) 199.63
MDL Number MFCD00077921
InChI Key CVZZNRXMDCOHBG-QMMMGPOBSA-N
Synonym 2-chloro-l-phenylalanine, l-2-chlorophenylalanine, s-2-amino-3-2-chlorophenyl propanoic acid, h-phe 2-cl-oh, 2s-2-amino-3-2-chlorophenyl propanoic acid, phenylalanine, 2-chloro, l-2-chlorophe, l-2-chloro phenylalanine, s-2-chlorophenylalanine, ksc491q0l
PubChem CID 2761491
IUPAC Name (2S)-2-amino-3-(2-chlorophenyl)propanoic acid
SMILES N[C@@H](CC1=C(Cl)C=CC=C1)C(O)=O

Especificaciones

Melting Point 196°C to 199°C
Solubility Information Insoluble in water.
Quantity 250 mg
Formula Weight 199.64
Percent Purity ≥98%
Chemical Name or Material 2-Chloro-L-phenylalanine

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.