missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N-(2,3-dimethylphenyl)benzamide, 97%

Product Code. 15462668
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15462668 250 mg 250mg
15452668 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15462668 Supplier Thermo Scientific Alfa Aesar Supplier No. H56225.MD
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 196617-88-6 | C15H14ClNO | 259.733 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 196617-88-6
Molecular Formula C15H14ClNO
Molecular Weight (g/mol) 259.733
MDL Number MFCD00018254
InChI Key OUAGJVUPQKYHDU-UHFFFAOYSA-N
Synonym 2-chloro-n-2,3-dimethylphenyl benzamide, cambridge id 5328590, 2-chloro-2',3'-benzoxylidide, 2-chloro-n-2,3-dimethyl-phenyl-benzamide, n-2,3-dimethylphenyl 2-chlorophenyl carboxamide
PubChem CID 295079
IUPAC Name 2-chloro-N-(2,3-dimethylphenyl)benzamide
SMILES CC1=C(C(=CC=C1)NC(=O)C2=CC=CC=C2Cl)C

Specifications

Quantity 250 mg
Formula Weight 259.74
Percent Purity 97%
Chemical Name or Material 2-Chloro-N-(2,3-dimethylphenyl)benzamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.